Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger GP Motor 3 HP 3 450 RPM 208-23

Catalog No. 502792553
Encompass_Preferred

GP Motor 3 HP 3 450 RPM 208-230V AC 56HC

Catalog No. 50-279-2553 Supplier Grainger Supplier No. 19YU90
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GP Motor 3 HP 3 450 RPM 208-230V AC 56HC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.