Learn More
Air Science Usa Air Science GP Plus! formula Carbon Filter use with all FUMEBOX ductless fume hoods, AP60-001, general purpose, organics, solvents, alcohols, odors
SDP
Supplier:  Air Science Usa AP60001
Encompass_Preferred
Air Science GP Plus! formula Carbon Filter use with FUMEBOX, AP60-001, for use with organics
Catalog No. NC1148873