Learn More
AnaSpec gp100 (25-33), human, 1mg
Supplier:  AnaSpec AS62589
Encompass
gp100 (2533), human[KVPRNQDWL] ; MW 1155.3; (1 mg) This is amino acids 25 to 33 fragment of human melanoma antigen gp100. This H-2Db restricted epitope is recognized by T cells. The gp100-specific, H-2Db-restricted, CD8+ T cells are capable of recognizing B16 melanoma but not normal melanocytes. This peptide was used as an immunogen in multiple cancer immunotherapy studies.
Catalog No. NC0888171