Learn More
Medchemexpress LLC HY-P1585A 1mg Medchemexpress, Gp100 (25-33), human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1585A1MG
Encompass_Preferred
Medchemexpress, HY-P1585A 1mg Gp100 (25-33), human (TFA) CAS: Gp100 (25-33), human TFA is the amino acids 25-33 fragment of the human melanoma antigen. It is a 9-amino acid (AA) epitope restricted by H-2D b and recognized by the T cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4231