Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6490MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: PMEL/783. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: SK-MEL-28 cells or Melanoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human SILV protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 90-100kDa.Chromosome: 12q13.2. Gene Symbol: SILV. Gene ID: 6490. Ensemble: ENSG00000185664. Uniprot: NP_001307051.
Catalog No. 50-188-5168