Learn More
ACROBiosystems ACROBiosystems HIV-1 [HIV-1/Clade C (16055)] GP120 Protein, His Tag
Catalog No. 501956757
Encompass_Preferred
Recombinant Protein;1MG;HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 (Accession # ABL67444.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.1 kDa. The protein migrates as 85-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 2 μg/mL (100 μL/well) can bind HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) with a linear range of 0.5-63 ng/mL (QC tested).
Catalog No. 50-195-6757 Supplier ACROBiosystems Supplier No. GP5V152241MG
Recombinant Protein;1MG;HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 (Accession # ABL67444.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.1 kDa. The protein migrates as 85-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 2 μg/mL (100 μL/well) can bind HIV-1 [HIV-1/Clade C (16055)] GP120, His Tag (Cat. No. GP5-V15224) with a linear range of 0.5-63 ng/mL (QC tested).