Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GP130 receptor agonist-1 | 339303-87-6 | 100.0% | 50 MG
SDP

Catalog No. 5000233665
Encompass_Preferred

GP130 receptor agonist-1 is a potent, brain-penetrant, orally active small-molecule agonist of the gp130 receptor with demonstrated neuroprotective activity against NMDA-induced neurotoxicity and modulation of STAT3, AKT, and ERK1/2 signaling in cellular and in vivo studies.

  • Potent brain-penetrant gp130 receptor agonist.
  • Orally active in preclinical dosing studies.
  • Demonstrated neuroprotective effect against NMDA-induced neurotoxicity.
  • Modulates STAT3, AKT, and ERK1/2 signaling pathways.
  • Molecular formula C15H11FN2S, molecular weight ~270.32 g/mol.
  • High purity (reported ~99.95%) and white to off-white solid appearance.
  • Soluble in DMSO; in vivo formulation protocols provided for suspension or oil-based dosing.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.

Catalog No. 50-002-33665 Supplier Medchemexpress LLC Supplier No. HY12148850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GP130 receptor agonist-1 is a potent, brain-penetrant, orally active small-molecule agonist of the gp130 receptor with demonstrated neuroprotective activity against NMDA-induced neurotoxicity and modulation of STAT3, AKT, and ERK1/2 signaling in cellular and in vivo studies.

  • Potent brain-penetrant gp130 receptor agonist.
  • Orally active in preclinical dosing studies.
  • Demonstrated neuroprotective effect against NMDA-induced neurotoxicity.
  • Modulates STAT3, AKT, and ERK1/2 signaling pathways.
  • Molecular formula C15H11FN2S, molecular weight ~270.32 g/mol.
  • High purity (reported ~99.95%) and white to off-white solid appearance.
  • Soluble in DMSO; in vivo formulation protocols provided for suspension or oil-based dosing.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.