Learn More
ACROBiosystems Recombinant Protein;Mouse gp130 / CD130 / IL-6 R beta Protein;HEK293;100UG;ILT,IL6ST,gp130,CD130,IL-6RB,IL-6R-beta,IL-6R subunit beta,IL-6 receptor subunit beta,CDw130,IL-6ST,IL6RB
Catalog No. 501658718
Encompass_Preferred
Recombinant Protein;Mouse gp130 / CD130 / IL-6 R beta Protein;100UG;Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Glu 617 (Accession # NP_034690).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 68.5 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8718 Supplier ACROBiosystems Supplier No. ILTM52H1100UG
Recombinant Protein;Mouse gp130 / CD130 / IL-6 R beta Protein;100UG;Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Glu 617 (Accession # NP_034690).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 68.5 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.