Learn More
ACROBiosystems gp140 200ug
Catalog No. 502534953
Encompass_Preferred
Biotinylated Human immunodeficiency virus 1 gp140 Protein HisAvitag (GP0-V82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Asp 661 (Accession # ABA61516.1).
Catalog No. 50-253-4953 Supplier ACROBiosystems Supplier No. GP0V82E3200UG
Biotinylated Human immunodeficiency virus 1 gp140 Protein HisAvitag (GP0-V82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Asp 661 (Accession # ABA61516.1).