Learn More
ACROBiosystems gp140 25ug
Supplier:  ACROBiosystems GP0V82E325UG
Encompass_Preferred
Biotinylated Human immunodeficiency virus 1 gp140 Protein HisAvitag (GP0-V82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Asp 661 (Accession # ABA61516.1).
Catalog No. 50-253-4952