Learn More
Aviva Systems Biology Corp GP1BA Antibody - C-terminal region (ARP51319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GP1BA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GP1BA. Alternate names: BSS, CD42B, CD42b-alpha, GP1B, MGC34595, VWDP, GPIbA, BDPLT1, BDPLT3, DBPLT3ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60544