Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2813MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: GP2/1712. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: PANC-1 cells. Pancreas. Cellular Staining: Cell Surface and Secreted. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human GP2 protein fragment (aa35-179). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 59kDa.Chromosome: 16p12. Gene Symbol: GP2. Gene ID: 2813. Ensemble: ENSG00000169347. Uniprot: NP_001007242.
Catalog No. 50-188-4749