Learn More
Aviva Systems Biology Corp GP6 Antibody - middle region (ARP47248_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47248_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GP6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GP6. Alternate names: GPIV, GPVI, MGC138168, BDPLT11ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59497