Learn More
Expression Systems gp64-PE (R-PHYCOERYTHRIN (R-PE)-CONJUGATED MOUSE ANTI-BACULOVIRUS MONOCLONAL ANTIBODY) 200ul

Supplier: Expression Systems 97201
Expression Systems offers a PE-labeled antibody directed to gp64, the baculovirus envelope protein required for cell-to-cell spread. The baculovirus gp64 envelope glycoprotein is a major component of the envelope of the budded baculovirus and is essential for entry into cultured insect host cells. The anti-gp64 antibody is effective for direct immunofluorescence staining of insect cells for flow cytometric analysis. Applications include baculovirus titration, infection kinetics monitoring and protein expression screening.
Contents:
gp64-PE (R-PHYCOERYTHRIN (R-PE)-CONJUGATED MOUSE ANTI-BACULOVIRUS MONOCLONAL ANTIBODY)
Clone: AcV1
Antibody Composition: Mouse IgG2A
Storage Buffer: Phosphate buffered saline (PBS) containing 0.05% BSA and 0.09% sodium azide.
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.