Learn More
Aviva Systems Biology Corp GP9 Antibody - middle region (ARP61533_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61533_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GP9. It was validated on Western Blot. . Alternate names: CD42a, GPIXApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-287-402