Learn More
ACROBiosystems Recombinant Protein;Mouse GPA33 / A33 Protein;HEK293;1MG;GP3,GPA33,A33,GP-A33
Catalog No. 501658086
Encompass_Preferred
Recombinant Protein;Mouse GPA33 / A33 Protein;1MG;Mouse GPA33, His Tag (GP3-M5225) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Ile 235 (Accession # NP_067623.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.7 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8086 Supplier ACROBiosystems Supplier No. GP3M52251MG
Recombinant Protein;Mouse GPA33 / A33 Protein;1MG;Mouse GPA33, His Tag (GP3-M5225) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Ile 235 (Accession # NP_067623.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.7 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.