Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human GPA33, ultra sensitivity (primary amine labeling);HEK293;200UG;GP3,GPA33,A33,GP-A33
Catalog No. 501659168
Encompass_Preferred
Recombinant Protein;Biotinylated Human GPA33, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human GPA33 / A33 (GP3-H8224) is expressed from human HEK293 cells. It contains AA Ile 22 - Val 235 (Accession # AAH74830). It is the biotinylated form of Human GPA33 / A33 Protein (Cat # GP3-H5224).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.5 kDa. The protein migrates as 35-40 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.
Catalog No. 50-165-9168 Supplier ACROBiosystems Supplier No. GP3H8224200UG
Recombinant Protein;Biotinylated Human GPA33, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human GPA33 / A33 (GP3-H8224) is expressed from human HEK293 cells. It contains AA Ile 22 - Val 235 (Accession # AAH74830). It is the biotinylated form of Human GPA33 / A33 Protein (Cat # GP3-H5224).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.5 kDa. The protein migrates as 35-40 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.