Learn More
ACROBiosystems Recombinant Protein;Human GPA33 / A33 Protein;HEK293;50UG;GP3,GPA33,A33,GP-A33
Catalog No. 501657891
Encompass_Preferred
Recombinant Protein;Human GPA33 / A33 Protein;50UG;Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # AAH74830).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.5 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7891 Supplier ACROBiosystems Supplier No. GP3H522450UG
Recombinant Protein;Human GPA33 / A33 Protein;50UG;Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # AAH74830).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.5 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.