Learn More
Medchemexpress LLC Gpat-In-1 | 1204347-51-2 | C21H26ClNO4S | 25 MG
SDP
Catalog No. 5000374690
Encompass_Preferred
GPAT-IN-1 is a glycerol-3-phosphate acyltransferase (GPAT) inhibitor with an IC50 value of 8.9 μM, making it suitable for research into obesity. It appears as a white to off-white solid with a molecular weight of 423.95 and a purity of 97.12%.
Catalog No. 50-003-74690 Supplier Medchemexpress LLC Supplier No. 25MGHY153628
GPAT-IN-1 is a glycerol-3-phosphate acyltransferase (GPAT) inhibitor with an IC50 value of 8.9 μM, making it suitable for research into obesity. It appears as a white to off-white solid with a molecular weight of 423.95 and a purity of 97.12%.