Learn More
Eurofins Discoverx cAMP Hunter GPBAR1 Bioassay Kit, 10-Plate. (95-0049Y2-00140)
Catalog No. 502002465
Encompass
The cAMP Hunter CHO-K1 GPBAR1 Bioassay Kit provides a robust and highly sensitive functional, cell-based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed to run the assay, including cryopreserved cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure long-term assay reproducibility, and faster implementation from characterization to QC lot release. This bioassay has been optimized for a 96-well format. The protocol can be optimized further for running the assay in a high-throughput 384-well format. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-2465 Supplier Eurofins Discoverx Supplier No. 950049Y200140
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter CHO-K1 GPBAR1 Bioassay Kit provides a robust and highly sensitive functional, cell-based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed to run the assay, including cryopreserved cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure long-term assay reproducibility, and faster implementation from characterization to QC lot release. This bioassay has been optimized for a 96-well format. The protocol can be optimized further for running the assay in a high-throughput 384-well format. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.