Learn More
Biomatik Corporation Gpc Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11895
Encompass_Preferred
Gpc Polyclonal Antibody is a Lymphocytic choriomeningitis virus (strain Armstrong) (LCMV) Polyclonal Antibody to GPC. The Gpc Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Lymphocytic choriomeningitis virus (strain Armstrong) (LCMV). Immunogen: Recombinant Lymphocytic Choriomeningitis Virus Pre-Glycoprotein Polyprotein Gp Complex Protein (266-498Aa). Applications: ELISA. Target Synonyms: GPC. Accession Number: P09991. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7615