Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp GPC Calibration Standard, 190,000 Da, Polystyrene, 100 mg

Catalog No. 502392126 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

An individual GPC standard (Gel Permeation Chromatography) for molecular weight determination. It is a Polystyrene Standard at 190,000 Da, 100 mg, (sample diluted with 100mL solvent yields 1mg/mL).

  • Individual GPC standard
  • For molecular weight determination
  • Polystyrene material
  • Molecular weight: 190,000 Da
  • Mass: 100 mg
  • Formulation: Solid
  • Storage condition: 5°C ± 3°C (Fridge)
  • Type: Calibration

Catalog No. 50-239-2126 Supplier Waters Corp Supplier No. 600000267
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An individual GPC standard (Gel Permeation Chromatography) for molecular weight determination. It is a Polystyrene Standard at 190,000 Da, 100 mg, (sample diluted with 100mL solvent yields 1mg/mL).

  • Individual GPC standard
  • For molecular weight determination
  • Polystyrene material
  • Molecular weight: 190,000 Da
  • Mass: 100 mg
  • Formulation: Solid
  • Storage condition: 5°C ± 3°C (Fridge)
  • Type: Calibration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.