Learn More
New England Biolabs, Inc. GpC Methyltransferase (M.CviPI) – 1000 units
SDP
Catalog No. 501526382
Encompass
Methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5´...GC...3´.
Catalog No. 50-152-6382 Supplier New England Biolabs, Inc. Supplier No. M0227L
Methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5´...GC...3´.