Learn More
AIRGAS GPC REGULATOR CGA-590 0-100PSI
Catalog No. NC2057588
Encompass
Airgas Two Stage Brass 0-100 psi General Purpose Cylinder Regulator CGA-590
Catalog No. NC2057588 Supplier AIRGAS Supplier No. Y12215D590AG
Airgas Two Stage Brass 0-100 psi General Purpose Cylinder Regulator CGA-590