Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2719MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: GPC3/863. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: 293T cells or Hepatocellular carcinoma. Cellular Staining: Cytoplasmic. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human GPC3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 67kDa.Chromosome: Xq26.2. Gene Symbol: GPC3. Gene ID: 2719. Ensemble: ENSG00000147257. Uniprot: NP_004475.
Catalog No. 50-188-4742