Learn More
ABclonal Technology GPC4 Rabbit pAb
SDP
Catalog No. 5000115913
Encompass_Preferred
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. The GPC4 gene is adjacent to the 3 end of GPC3 and may also play a role in Simpson-Golabi-Behmel syndrome.
Catalog No. 50-001-15913 Supplier ABclonal Technology Supplier No. A12805100UL
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. The GPC4 gene is adjacent to the 3 end of GPC3 and may also play a role in Simpson-Golabi-Behmel syndrome.