Learn More
Montana Molecular GPCR Kinase 2 Expression Kit - BacMam, 10mL - Montana Molecular
SDP
Catalog No. NC2600465
Encompass_Preferred
GPCR Kinase 2 (GRK2) packaged in BacMam for expression in mammalian cells. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity, enabling rapid assay optimization. Co-express with other Montana Molecular tools includeing GPCRs packaged in BacMam or biosensors for cAMP, DAG, or arrestin signaling.
Catalog No. NC2600465 Supplier Montana Molecular Supplier No. K0002N010
GPCR Kinase 2 (GRK2) packaged in BacMam for expression in mammalian cells. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity, enabling rapid assay optimization. Co-express with other Montana Molecular tools includeing GPCRs packaged in BacMam or biosensors for cAMP, DAG, or arrestin signaling.