Learn More
Montana Molecular GPCR Kinase 5 Expression Kit - BacMam, 10mL - Montana Molecular
SDP
Supplier:  Montana Molecular K0005N010
Encompass_Preferred
GPCR Kinase 5 (GRK5) packaged in BacMam for expression in mammalian cells. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity, enabling rapid assay optimization. Co-express with other Montana Molecular tools includeing GPCRs packaged in BacMam or biosensors for cAMP, DAG, or arrestin signaling.
Catalog No. NC2600472