Learn More
Aviva Systems Biology Corp GPD1 Antibody - middle region (ARP42228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GPD1. Alternate names: FLJ26652, HTGTIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58152