Learn More
Aviva Systems Biology Corp GPER Antibody - C-terminal region (ARP62243_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62243_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPER. It was validated on Western Blot. . Alternate names: CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPR30, LERGU, LERGU2, LyGPR, MGC99678, GPERApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-626