Learn More
TARGETMOL CHEMICALS INC GPI-1046 5MG
SDP
Catalog No. 502879464
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.GPI-1046 is an immunophilin ligand without antibiotic action and attenuates ethanol intake in part through the upregulation of glutamate transporter 1 (GLT1) in PFC and NAc-core. Purity 99.91%
Catalog No. 50-287-9464 Supplier TARGETMOL CHEMICALS INC Supplier No. T114515MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.GPI-1046 is an immunophilin ligand without antibiotic action and attenuates ethanol intake in part through the upregulation of glutamate transporter 1 (GLT1) in PFC and NAc-core. Purity 99.91%