Learn More
Medchemexpress LLC HY-136424 5mg Medchemexpress, GPI-1485 CAS:186268-78-0 Purity:>98%
SDP
Catalog No. 502030437
Encompass_Preferred
Medchemexpress, HY-136424 5mg GPI-1485 CAS:186268-78-0 GPI-1485 (GM1485), a nonimmunosuppressive immunophilin ligand, promotes neurofunctional improvement and neural regeneration following stroke. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0437 Supplier Medchemexpress LLC Supplier No. HY1364245MG
Medchemexpress, HY-136424 5mg GPI-1485 CAS:186268-78-0 GPI-1485 (GM1485), a nonimmunosuppressive immunophilin ligand, promotes neurofunctional improvement and neural regeneration following stroke. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.