Learn More
Medchemexpress LLC GPI-1485 (GM1485) | 186268-78-0 | 99.6% | 241.28 | 1 ML
SDP
Catalog No. 5000351167
Encompass_Preferred
GPI-1485 (GM1485) is a nonimmunosuppressive immunophilin ligand that promotes neurofunctional improvement and neural regeneration after a stroke. It is intended for research use only.
  • Promotes neurofunctional improvement and neural regeneration
  • Acts as a nonimmunosuppressive immunophilin ligand
  • Induces cellular reprogramming and neural cell induction in vitro
  • Improves long-term neurological recovery in vivo
  • Supplied as a white to light yellow solid
Catalog No. 50-003-51167 Supplier Medchemexpress LLC Supplier No. HY13642410MM/1ML
Add to Cart
Add to Cart
GPI-1485 (GM1485) is a nonimmunosuppressive immunophilin ligand that promotes neurofunctional improvement and neural regeneration after a stroke. It is intended for research use only.
  • Promotes neurofunctional improvement and neural regeneration
  • Acts as a nonimmunosuppressive immunophilin ligand
  • Induces cellular reprogramming and neural cell induction in vitro
  • Improves long-term neurological recovery in vivo
  • Supplied as a white to light yellow solid