Learn More
Medchemexpress LLC Gpi-1485 (gm1485) | 186268-78-0 | 99.6% | 241.28 | 250 MG
SDP
Catalog No. 5000348784
Encompass_Preferred
GPI-1485 (GM1485) is a nonimmunosuppressive immunophilin ligand that promotes neurofunctional improvement and neural regeneration following stroke.
  • Targets FKBP
  • Induces cellular reprogramming and neural cell induction
  • Improves long-term neurological recovery in animal models
  • Available as a white to light yellow solid
  • Stable for up to 3 years as powder at -20°C
  • Stable for up to 2 years in solvent at -80°C
Catalog No. 50-003-48784 Supplier Medchemexpress LLC Supplier No. 250MGHY136424
GPI-1485 (GM1485) is a nonimmunosuppressive immunophilin ligand that promotes neurofunctional improvement and neural regeneration following stroke.
  • Targets FKBP
  • Induces cellular reprogramming and neural cell induction
  • Improves long-term neurological recovery in animal models
  • Available as a white to light yellow solid
  • Stable for up to 3 years as powder at -20°C
  • Stable for up to 2 years in solvent at -80°C