Learn More
Ace Glass, Inc. PTFE connecting adapter multipack with Chemraz O-rings, connects 15mm Ace-Thred to 8-425, 13-425, 15-425, 18-400, 20-400, 22-400 and 24-410 vial threads. Also includes one13290-34 glass adapter.
SDP
Catalog No. 50977944
Encompass
PTFE connecting adapter multipack with Chemraz O-rings, connects 15mm Ace-Thred to 8-425, 13-425, 15-425, 18-400, 20-400, 22-400 and 24-410 vial threads. Also includes one13290-34 glass adapter.
Catalog No. 50-977-944 Supplier Ace Glass, Inc. Supplier No. 1329059
PTFE connecting adapter multipack with Chemraz O-rings, connects 15mm Ace-Thred to 8-425, 13-425, 15-425, 18-400, 20-400, 22-400 and 24-410 vial threads. Also includes one13290-34 glass adapter.