Learn More
Ace Glass, Inc. PTFE connecting adapter multipack with Chemraz O-rings, connects 15mm Ace-Thred to 8-425, 13-425, 15-425, 18-400, 20-400, 22-400 and 24-410 vial threads. Also includes one13290-45 glass adapter.
SDP
Catalog No. 50983487
Encompass
PTFE connecting adapter multipack with Chemraz O-rings, connects 15mm Ace-Thred to 8-425, 13-425, 15-425, 18-400, 20-400, 22-400 and 24-410 vial threads. Also includes one13290-45 glass adapter.
Catalog No. 50-983-487 Supplier Ace Glass, Inc. Supplier No. 1329069
PTFE connecting adapter multipack with Chemraz O-rings, connects 15mm Ace-Thred to 8-425, 13-425, 15-425, 18-400, 20-400, 22-400 and 24-410 vial threads. Also includes one13290-45 glass adapter.