Learn More
Aviva Systems Biology Corp GPI Antibody - C-terminal region (ARP61088_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61088_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPI. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GPI. Alternate names: AMF, GNPI, NLK, PGI, PHI, SA36, SA-36ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-287-245