Learn More
Aviva Systems Biology Corp GPIHBP1 Antibody - C-terminal region (ARP64479_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64479_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPIHBP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human GPIHBP1. Alternate names: GPI-HBP1ApplicationWB. Species reactivity: Human
Catalog No. 50-288-271