Learn More
Medchemexpress LLC GPLGIAGQ TFA | 99.9% | 825.83 | 5 MG
SDP
Catalog No. 5000424155
Encompass_Preferred
GPLGIAGQ TFA is a polypeptide that can be cleaved by Matrix Metalloproteinase 2 (MMP2). It is utilized as a stimulus-sensitive linker in liposomal and micellar nanocarriers for tumor targeting that is triggered by MMP2. This polypeptide can also be used in the synthesis of unique MMP2-targeted photosensitizers for photodynamic therapy (PDT).
  • MMP2-cleavable polypeptide.
  • Used as a stimulus-sensitive linker in liposomal and micellar nanocarriers.
  • Enables MMP2-triggered tumor targeting.
  • Can be used to synthesize MMP2-targeted photosensitizers in photodynamic therapy (PDT).
  • Triggers PEG deshielding of liposomal carriers, enhancing cellular internalization.
Catalog No. 50-004-24155 Supplier Medchemexpress LLC Supplier No. HYP2213A5MG
GPLGIAGQ TFA is a polypeptide that can be cleaved by Matrix Metalloproteinase 2 (MMP2). It is utilized as a stimulus-sensitive linker in liposomal and micellar nanocarriers for tumor targeting that is triggered by MMP2. This polypeptide can also be used in the synthesis of unique MMP2-targeted photosensitizers for photodynamic therapy (PDT).
  • MMP2-cleavable polypeptide.
  • Used as a stimulus-sensitive linker in liposomal and micellar nanocarriers.
  • Enables MMP2-triggered tumor targeting.
  • Can be used to synthesize MMP2-targeted photosensitizers in photodynamic therapy (PDT).
  • Triggers PEG deshielding of liposomal carriers, enhancing cellular internalization.