Learn More
Aviva Systems Biology Corp GPN3 Antibody - C-terminal region (ARP62840_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62840_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPN3. It was validated on Western Blot. . Alternate names: ATPBD1C, MGC14560, MGC32810ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-783