Learn More
Medchemexpress LLC HY-W011391 10mg Medchemexpress, GPNA (hydrochloride) CAS:67953-08-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01139110MG
Encompass_Preferred
Medchemexpress, HY-W011391 10mg GPNA (hydrochloride) CAS:67953-08-6 GPNA hydrochloride is a well known substrate of the enzyme γ-glutamyltransferase (GGT). GPNA hydrochloride is a specific glutamine (Gln) transporter ASCT2 inhibitor. GPNA hydrochloride also inhibit Na+-dependent carriers, such as SNAT family (SNAT1/2/4/5), and the Na+-independent leucine transporters LAT1/2. GPNA reversibly induces apoptosis in A549 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3312