Learn More
Selleck Chemical LLC GPNA hydrochloride
SDP
Supplier:  Selleck Chemical LLC S6670100MG
Encompass_Preferred
GPNA hydrochloride (L- -glutamyl-p-nitroanilide hydrochloride) is a substrate of -glutamyltransferase (GGT) and an inhibitor of the glutamine transporter ASCT2 with an IC50 of 250 M in A549 cells It also inhibits Na -dependent carriers SNAT1 -2 -4 -5 and Na -independent leucine transporters LAT1/2 It can be widely used in research on pancreatic cancer breast cancer glioblastoma multiforme acute myelogenous leukemia and non-small-cell lung cancer
Catalog No. 50-000-02675