Learn More
TARGETMOL CHEMICALS INC GPNA HYDROCHLORIDE 50MG
SDP
Catalog No. 502577355
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GPNA hydrochloride is specific glutamine (Gln) transporter ASCT2(SLC1A5) inhibitor. GPNA reversibly induces apoptosis in A549 cells. GPNA hydrochloride is a well-known substrate of the enzyme (gamma)-glutamyltransferase (GGT). GPNA hydrochloride also inhibits Na+-dependent carriers such as SNAT family (SNAT1/2/4/5) and the Na+-independent leucine transporters LAT1/2. purity: 99%
Catalog No. 50-257-7355 Supplier TARGETMOL CHEMICALS INC Supplier No. T1541150MG
Also available in 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GPNA hydrochloride is specific glutamine (Gln) transporter ASCT2(SLC1A5) inhibitor. GPNA reversibly induces apoptosis in A549 cells. GPNA hydrochloride is a well-known substrate of the enzyme (gamma)-glutamyltransferase (GGT). GPNA hydrochloride also inhibits Na+-dependent carriers such as SNAT family (SNAT1/2/4/5) and the Na+-independent leucine transporters LAT1/2. purity: 99%