Learn More
Medchemexpress LLC GPNA hydrochloride | 67953-08-6 | 99.6% | 303.70 | 1 ML
SDP
Catalog No. 5000455013
Encompass_Preferred
GPNA hydrochloride is identified as a substrate of the enzyme γ-glutamyltransferase (GGT) and a specific glutamine (Gln) transporter ASCT2 inhibitor. It also inhibits Na+-dependent carriers like the SNAT family (SNAT1/2/4/5) and Na+-independent leucine transporters LAT1/2. GPNA reversibly induces apoptosis in A549 cells.
  • Substrate of the enzyme γ-glutamyltransferase (GGT)
  • Specific glutamine (Gln) transporter ASCT2 inhibitor
  • Inhibits Na+-dependent carriers (SNAT family)
  • Inhibits Na+-independent leucine transporters (LAT1/2)
  • Reversibly induces apoptosis in A549 cells
Catalog No. 50-004-55013 Supplier Medchemexpress LLC Supplier No. HYW01139110MM/1ML
GPNA hydrochloride is identified as a substrate of the enzyme γ-glutamyltransferase (GGT) and a specific glutamine (Gln) transporter ASCT2 inhibitor. It also inhibits Na+-dependent carriers like the SNAT family (SNAT1/2/4/5) and Na+-independent leucine transporters LAT1/2. GPNA reversibly induces apoptosis in A549 cells.
  • Substrate of the enzyme γ-glutamyltransferase (GGT)
  • Specific glutamine (Gln) transporter ASCT2 inhibitor
  • Inhibits Na+-dependent carriers (SNAT family)
  • Inhibits Na+-independent leucine transporters (LAT1/2)
  • Reversibly induces apoptosis in A549 cells