Learn More
Aviva Systems Biology Corp GPNMB Antibody - N-terminal region (ARP44499_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44499_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPNMB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GPNMB. Alternate names: HGFIN, NMBApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58826