Learn More
Medchemexpress LLC GPNMB/Osteoactivin 5ug
SDP
Catalog No. 5000197161
Encompass_Preferred
Recombinant human GPNMB (Osteoactivin), expressed in HEK293 cells with a C-terminal His tag, is supplied as a lyophilized, glycosylated protein of >95% purity and is functionally active in cell-based assays. It is intended for in vitro receptor binding, signaling studies, and assay development; aliquot and store frozen after reconstitution to preserve activity.
  • High purity glycosylated protein suitable for functional assays.
  • Expressed in mammalian cells for native post-translational modifications.
  • Lyophilized format for long-term storage and transport.
  • Validated activity in cell proliferation assays.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.
Catalog No. 50-001-97161 Supplier Medchemexpress LLC Supplier No. HYP708365UG
Recombinant human GPNMB (Osteoactivin), expressed in HEK293 cells with a C-terminal His tag, is supplied as a lyophilized, glycosylated protein of >95% purity and is functionally active in cell-based assays. It is intended for in vitro receptor binding, signaling studies, and assay development; aliquot and store frozen after reconstitution to preserve activity.
  • High purity glycosylated protein suitable for functional assays.
  • Expressed in mammalian cells for native post-translational modifications.
  • Lyophilized format for long-term storage and transport.
  • Validated activity in cell proliferation assays.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.