Learn More
Abcam GppCp (GMPPCP), Nonhydrolyzable GTP analog, 25MG
Supplier:  Abcam AB14666025MG
Encompass_Preferred
MW 521.21 g/mol, Purity >95%. Nonhydrolyzable GTP analog. Modulates G-protein cell signaling, protein synthesis and other metabolic processes. Forms stable protein complexes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1666769
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.