Learn More
Eurofins Discoverx Ready-to-Assay GPR103/Peptide P518 Receptor Frozen Cells, 2 Vials of Cells. (HTS189RTA)
Catalog No. 502006036
Encompass
The Ready-to-Assay GPR103/Peptide P518 Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. QRFP neuropeptides) based activation of QRFPR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses QRFPR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of QRFPR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-6036 Supplier Eurofins Discoverx Supplier No. HTS189RTA
The Ready-to-Assay GPR103/Peptide P518 Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. QRFP neuropeptides) based activation of QRFPR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses QRFPR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of QRFPR, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.