Learn More
Aviva Systems Biology Corp GPR12 Antibody - C-terminal region (ARP64695_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64695_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPR12. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human GPR12. Alternate names: FLJ18149, FLJ97704, GPCR12, GPCR21, MGC138349, MGC138351ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-338