Learn More
Cayman Chemical GPR120 Compound A 10mg
SDP
Catalog No. 502980390
Encompass_Preferred
An orally available, selective agonist of GPR120 (EC50 = ~0.35 µM); exerts anti-inflammatory effects on macrophages in vitro and improves glucose tolerance, decreases hyperinsulinemia, increases insulin sensitivity, and decreases hepatic steatosis when included at 30 mg/kg body weight in a high-fat diet fed to obese mice
Catalog No. 50-298-0390 Supplier Cayman Chemical Supplier No. 1662410 MG
An orally available, selective agonist of GPR120 (EC50 = ~0.35 µM); exerts anti-inflammatory effects on macrophages in vitro and improves glucose tolerance, decreases hyperinsulinemia, increases insulin sensitivity, and decreases hepatic steatosis when included at 30 mg/kg body weight in a high-fat diet fed to obese mice